Assay Details
| Gene Symbol: AVP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213952.2 | NP_999117.1 | 1 | 152 | 86 | |
| GenBank mRNA | M25644.1 | - | 1 | 153 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: AVP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213952.2 | NP_999117.1 | 1 | 152 | 86 | |
| GenBank mRNA | M25644.1 | - | 1 | 153 | 86 | |
Set Membership: |
|