Assay Details
| Gene Symbol: HPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213953.2 | NP_999118.1 | 10 | 1251 | 50 | |
| XM_021102207.1 | XP_020957866.1 | 1278 | 50 | |||
| GenBank mRNA | AK232392.1 | - | 10 | 1253 | 50 | |
| AK232781.1 | - | 10 | 1259 | 50 | ||
| AK393501.1 | - | 10 | 1367 | 50 | ||
| AK394238.1 | - | 8 | 927 | 50 | ||
| AK394246.1 | - | 10 | 1254 | 50 | ||
| U14751.1 | - | 10 | 1254 | 50 | ||
More Information
Set Membership: |
|

HPX
Genomic Map