Assay Details
| Gene Symbol: TDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214004.1 | NP_999169.1 | 8 | 1046 | 103 | |
| XM_005658621.3 | XP_005658678.1 | 1160 | 103 | |||
| XM_021072090.1 | XP_020927749.1 | 1248 | 103 | |||
| XM_021072091.1 | XP_020927750.1 | 1385 | 103 | |||
| XM_021072092.1 | XP_020927751.1 | 1192 | 103 | |||
| XM_021072093.1 | XP_020927752.1 | 1128 | 103 | |||
| GenBank mRNA | AK233173.1 | - | 10 | 1247 | 103 | |
| AK236583.1 | - | 8 | 1254 | 103 | ||
| AK400666.1 | - | 10 | 1249 | 103 | ||
| AY095535.1 | - | 8 | 1046 | 103 | ||
More Information
Set Membership: |
|

TDH
Genomic Map