Assay Details
Gene Symbol: AMBN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214037.1 | NP_999202.1 | 11 | 1246 | 98 | |
GenBank mRNA | AK231301.1 | - | 5 | 786 | 98 | |
U43404.1 | - | 11 | 1246 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
