Assay Details
| Gene Symbol: ACADM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214039.1 | NP_999204.1 | 11 | 1144 | 127 | |
| GenBank mRNA | AK230498.1 | - | 11 | 1195 | 127 | |
| AK235593.1 | - | 11 | 1192 | 127 | ||
| AK236293.1 | - | 11 | 1161 | 127 | ||
| AY705916.1 | - | 11 | 1134 | 127 | ||
| U40845.1 | - | 11 | 1144 | 127 | ||
More Information
Set Membership: |
|

ACADM
Genomic Map