Assay Details
| Gene Symbol: HMGB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214063.1 | NP_999228.1 | 3 - 4 | 445 | 63 | |
| XM_021072104.1 | XP_020927763.1 | 478 | 63 | |||
| XM_021072105.1 | XP_020927764.1 | 445 | 63 | |||
| GenBank mRNA | AK234412.1 | - | 3 - 4 | 447 | 63 | |
| AK238980.1 | - | 3 - 4 | 470 | 63 | ||
| AK394538.1 | - | 3 - 4 | 499 | 63 | ||
| AK399703.1 | - | 3 - 4 | 513 | 63 | ||
| AY609856.1 | - | 3 - 4 | 459 | 63 | ||
| J02895.1 | - | 3 - 4 | 445 | 63 | ||
More Information
Set Membership: |
|

HMGB2
Genomic Map