Assay Details
| Gene Symbol: HMGB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214063.1 | NP_999228.1 | 4 - 5 | 615 | 85 | |
| XM_021072104.1 | XP_020927763.1 | 648 | 85 | |||
| XM_021072105.1 | XP_020927764.1 | 615 | 85 | |||
| GenBank mRNA | AK234412.1 | - | 4 - 5 | 617 | 85 | |
| AK238980.1 | - | 4 - 5 | 640 | 85 | ||
| AK394538.1 | - | 4 - 5 | 669 | 85 | ||
| AY609856.1 | - | 4 - 5 | 629 | 85 | ||
| J02895.1 | - | 4 - 5 | 615 | 85 | ||
More Information
Set Membership: |
|

HMGB2
Genomic Map