Assay Details
| Gene Symbol: FUT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214069.1 | NP_999234.1 | 2 | 448 | 111 | |
| GenBank mRNA | AF136895.1 | - | 2 | 448 | 111 | |
| AK239893.1 | - | 2 | 568 | 111 | ||
| AK396605.1 | - | 2 | 454 | 111 | ||
| Gene Symbol: FUT2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003127283.4 | XP_003127331.1 | 658 | 111 | ||
| XM_013998595.2 | XP_013854049.1 | 670 | 111 | |||
| XM_013998597.2 | XP_013854051.1 | 681 | 111 | |||
| XM_013998598.2 | XP_013854052.1 | 665 | 111 | |||
| XM_021094686.1 | XP_020950345.1 | 683 | 111 | |||
| GenBank mRNA | AK350464.1 | - | 4 | 658 | 111 | |
More Information
Set Membership: |
|

FUT2
Genomic Map