Assay Details
| Gene Symbol: HTR7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214085.1 | NP_999250.1 | 1 - 2 | 537 | 71 | |
| XM_005671276.3 | XP_005671333.1 | 638 | 71 | |||
| XM_021072106.1 | XP_020927765.1 | 638 | 71 | |||
| GenBank mRNA | AF420595.1 | - | 1 - 2 | 537 | 71 | |
More Information
Set Membership: |
|

HTR7
Genomic Map