Assay Details
| Gene Symbol: OPN1SW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214090.1 | NP_999255.1 | 1 - 2 | 365 | 78 | |
| GenBank mRNA | AY091587.1 | - | 1 - 2 | 365 | 78 | |
More Information
Set Membership: |
|

OPN1SW
Genomic Map