Assay Details
| Gene Symbol: CD69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214091.2 | NP_999256.1 | 2 - 3 | 191 | 90 | |
| GenBank mRNA | AF484233.1 | - | 2 - 3 | 192 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CD69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214091.2 | NP_999256.1 | 2 - 3 | 191 | 90 | |
| GenBank mRNA | AF484233.1 | - | 2 - 3 | 192 | 90 | |
Set Membership: |
|