Assay Details
| Gene Symbol: CD69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214091.2 | NP_999256.1 | 4 - 5 | 506 | 89 | |
| GenBank mRNA | AF484233.1 | - | 4 - 5 | 507 | 89 | |
| AF484234.1 | - | 3 - 4 | 384 | 89 | ||
More Information
Set Membership: |
|

CD69
Genomic Map