Assay Details
| Gene Symbol: HSP90B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214103.1 | NP_999268.1 | 14 - 15 | 2040 | 86 | |
| XM_021091129.1 | XP_020946788.1 | 2191 | 86 | |||
| GenBank mRNA | AK236878.1 | - | 14 - 15 | 2064 | 86 | |
| AK390920.1 | - | 14 - 15 | 2135 | 86 | ||
| X76301.1 | - | 2028 | 86 | |||
| Y09136.1 | - | 14 - 15 | 2040 | 86 | ||
More Information
Set Membership: |
|

HSP90B1
Genomic Map