Assay Details
| Gene Symbol: HAMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214117.1 | NP_999282.1 | 1 - 2 | 146 | 54 | |
| GenBank mRNA | AF516143.1 | - | 1 - 2 | 146 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: HAMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214117.1 | NP_999282.1 | 1 - 2 | 146 | 54 | |
| GenBank mRNA | AF516143.1 | - | 1 - 2 | 146 | 54 | |
Set Membership: |
|