Assay Details
| Gene Symbol: IL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214135.2 | NP_999300.1 | 4 - 5 | 584 | 64 | |
| XM_013996658.2 | XP_013852112.1 | 7724 | 64 | |||
| GenBank mRNA | AB035380.2 | - | 4 - 5 | 584 | 64 | |
| AB040441.1 | - | 3 - 4 | 355 | 64 | ||
| AK351694.1 | - | 4 - 5 | 580 | 64 | ||
| EU364895.1 | - | 3 - 4 | 355 | 64 | ||
| KF246514.1 | - | 4 - 5 | 378 | 64 | ||
More Information
Set Membership: |
|

IL7
Genomic Map