Assay Details
| Gene Symbol: CLCN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214139.1 | NP_999304.1 | 5 - 6 | 784 | 100 | |
| GenBank mRNA | AF274055.1 | - | 5 - 6 | 784 | 100 | |
More Information
Set Membership: |
|

CLCN5
Genomic Map