Assay Details
| Gene Symbol: CD59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214170.1 | NP_999335.1 | 2 - 3 | 157 | 69 | |
| XM_005661031.3 | XP_005661088.1 | 305 | 69 | |||
| GenBank mRNA | AF020302.1 | - | 2 - 3 | 157 | 69 | |
| AK238416.1 | - | 2 - 3 | 121 | 69 | ||
More Information
Set Membership: |
|

CD59
Genomic Map