Assay Details
| Gene Symbol: NR5A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214179.1 | NP_999344.1 | 3 - 4 | 1736 | 59 | |
| XM_021082954.1 | XP_020938613.1 | 1162 | 59 | |||
| XM_021082957.1 | XP_020938616.1 | 939 | 59 | |||
| GenBank mRNA | U84399.1 | - | 3 - 4 | 1736 | 59 | |
More Information
Set Membership: |
|

NR5A1
Genomic Map