Assay Details
| Gene Symbol: AMY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214195.1 | NP_999360.1 | 3 - 4 | 512 | 75 | |
| XM_005663621.3 | XP_005663678.2 | 735 | 75 | |||
| XM_005663623.3 | XP_005663680.2 | 821 | 75 | |||
| XM_021088708.1 | XP_020944367.1 | 817 | 75 | |||
| XM_021088709.1 | XP_020944368.1 | 941 | 75 | |||
| XM_021088710.1 | XP_020944369.1 | 634 | 75 | |||
| XM_021088711.1 | XP_020944370.1 | 577 | 75 | |||
| GenBank mRNA | AF064742.1 | - | 3 - 4 | 512 | 75 | |
| AK395394.1 | - | 4 - 5 | 673 | 75 | ||
| Gene Symbol: LOC100521789 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003125881.6 | XP_003125929.2 | 863 | 75 | ||
| XM_005663625.3 | XP_005663682.2 | 695 | 75 | |||
| XM_021090122.1 | XP_020945781.1 | 801 | 75 | |||
| XM_021090123.1 | XP_020945782.1 | 851 | 75 | |||
| XM_021090124.1 | XP_020945783.1 | 577 | 75 | |||
More Information
Set Membership: |
|

AMY2
Genomic Map