Assay Details
| Gene Symbol: TGFB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214198.1 | NP_999363.1 | 4 - 5 | 1016 | 104 | |
| XM_005666355.3 | XP_005666412.1 | 2066 | 104 | |||
| XM_013978322.2 | XP_013833776.1 | 1796 | 104 | |||
| GenBank mRNA | AK236343.1 | - | 5 - 6 | 2047 | 104 | |
| AK240494.1 | - | 4 - 5 | 466 | 104 | ||
| X14150.1 | - | 1016 | 104 | |||
More Information
Set Membership: |
|

TGFB3
Genomic Map