Assay Details
| Gene Symbol: PKIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214204.1 | NP_999369.1 | 2 - 3 | 199 | 101 | |
| GenBank mRNA | AF132737.1 | - | 2 - 3 | 199 | 101 | |
| AK238759.1 | - | 3 - 4 | 456 | 101 | ||
| AK389967.1 | - | 3 - 4 | 456 | 101 | ||
More Information
Set Membership: |
|

PKIA
Genomic Map