Assay Details
| Gene Symbol: FABP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214215.2 | NP_999380.2 | 1 - 2 | 141 | 73 | |
| GenBank mRNA | AK231219.1 | - | 1 - 2 | 129 | 73 | |
| BW983662.1 | - | 1 - 2 | 128 | 73 | ||
| EW435209.2 | - | 1 - 2 | 167 | 73 | ||
| J05136.1 | - | 2 - 3 | 171 | 73 | ||
More Information
Set Membership: |
|

FABP6
Genomic Map