Assay Details
| Gene Symbol: RHO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214221.1 | NP_999386.1 | 4 - 5 | 931 | 68 | |
| GenBank mRNA | AF008947.1 | - | 4 - 5 | 931 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: RHO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214221.1 | NP_999386.1 | 4 - 5 | 931 | 68 | |
| GenBank mRNA | AF008947.1 | - | 4 - 5 | 931 | 68 | |
Set Membership: |
|