Assay Details
| Gene Symbol: CD86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214222.1 | NP_999387.1 | 2 - 3 | 384 | 126 | |
| XM_005654040.3 | XP_005654097.1 | 126 | ||||
| XM_005654041.3 | XP_005654098.1 | 126 | ||||
| XM_005654042.3 | XP_005654099.1 | 126 | ||||
| XM_005654043.3 | XP_005654100.1 | 126 | ||||
| XM_005654044.3 | XP_005654101.1 | 126 | ||||
| XM_005654045.3 | XP_005654102.1 | 126 | ||||
| XM_005654046.3 | XP_005654103.1 | 126 | ||||
| GenBank mRNA | AK231046.1 | - | 3 - 4 | 126 | ||
| AK343921.1 | - | 3 - 4 | 126 | |||
| AK391252.1 | - | 3 - 4 | 126 | |||
| AK399465.1 | - | 2 - 3 | 126 | |||
| AY826403.1 | - | 2 - 3 | 126 | |||
| AY834754.1 | - | 2 - 3 | 126 | |||
| KF646138.1 | - | 2 - 3 | 126 | |||
| L76099.1 | - | 2 - 3 | 384 | 126 | ||
More Information
Set Membership: |
|

CD86
Genomic Map