Assay Details
| Gene Symbol: CPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214244.1 | NP_999409.1 | 4 - 5 | 487 | 52 | |
| XM_005673165.3 | XP_005673222.1 | 1024 | 52 | |||
| GenBank mRNA | AF076222.1 | - | 4 - 5 | 487 | 52 | |
More Information
Set Membership: |
|

CPA1
Genomic Map