Assay Details
| Gene Symbol: ODF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214255.2 | NP_999420.1 | 1 - 2 | 441 | 81 | |
| GenBank mRNA | X69513.1 | - | 441 | 81 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: ODF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214255.2 | NP_999420.1 | 1 - 2 | 441 | 81 | |
| GenBank mRNA | X69513.1 | - | 441 | 81 | ||
Set Membership: |
|