Assay Details
| Gene Symbol: CS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214276.1 | NP_999441.1 | 7 - 8 | 790 | 61 | |
| XM_021091143.1 | XP_020946802.1 | 919 | 61 | |||
| XM_021091144.1 | XP_020946803.1 | 1075 | 61 | |||
| XM_021091145.1 | XP_020946804.1 | 1060 | 61 | |||
| XM_021091147.1 | XP_020946806.1 | 603 | 61 | |||
| GenBank mRNA | AK231644.1 | - | 7 - 8 | 972 | 61 | |
| AK235732.1 | - | 8 - 9 | 1004 | 61 | ||
| AK236734.1 | - | 8 - 9 | 1003 | 61 | ||
| AK238572.1 | - | 8 - 9 | 1002 | 61 | ||
| AK389252.1 | - | 6 - 7 | 670 | 61 | ||
| AK389361.1 | - | 7 - 8 | 967 | 61 | ||
| AK390176.1 | - | 7 - 8 | 1013 | 61 | ||
| AK394047.1 | - | 8 - 9 | 1005 | 61 | ||
| AK395560.1 | - | 8 - 9 | 1022 | 61 | ||
| AK396408.1 | - | 7 - 8 | 910 | 61 | ||
| AK399051.1 | - | 1315 | 61 | |||
| AK400607.1 | - | 7 - 8 | 932 | 61 | ||
| AK400978.1 | - | 7 - 8 | 927 | 61 | ||
| DQ403143.1 | - | 1 - 2 | 118 | 61 | ||
| M21197.1 | - | 7 - 8 | 790 | 61 | ||
More Information
Set Membership: |
|

CS
Genomic Map