Assay Details
| Gene Symbol: RND3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214296.1 | NP_999461.1 | 3 - 4 | 347 | 82 | |
| GenBank mRNA | AF078839.1 | - | 3 - 4 | 347 | 82 | |
| AK389995.1 | - | 3 - 4 | 577 | 82 | ||
| AY610204.1 | - | 3 - 4 | 562 | 82 | ||
More Information
Set Membership: |
|

RND3
Genomic Map