Assay Details
| Gene Symbol: APOE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214308.1 | NP_999473.1 | 2 - 3 | 96 | 109 | |
| GenBank mRNA | AK232315.1 | - | 2 - 3 | 116 | 109 | |
| AK399873.1 | - | 2 - 3 | 117 | 109 | ||
| X72835.1 | - | 96 | 109 | |||
More Information
Set Membership: |
|

APOE
Genomic Map