Assay Details
| Gene Symbol: TPM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214335.1 | NP_999500.1 | 3 - 4 | 389 | 57 | |
| XM_005661198.1 | XP_005661255.1 | 688 | 57 | |||
| XM_021081566.1 | XP_020937225.1 | 261 | 57 | |||
| GenBank mRNA | AB530159.1 | - | 3 - 4 | 389 | 57 | |
| AF087679.1 | - | 3 - 4 | 389 | 57 | ||
| AK232365.1 | - | 3 - 4 | 488 | 57 | ||
| AK233008.1 | - | 4 - 5 | 563 | 57 | ||
| AK234839.1 | - | 5 - 6 | 563 | 57 | ||
| AK235061.1 | - | 3 - 4 | 513 | 57 | ||
| AK235278.1 | - | 4 - 5 | 567 | 57 | ||
| AK235346.1 | - | 4 - 5 | 568 | 57 | ||
| AK235570.1 | - | 4 - 5 | 563 | 57 | ||
| AK235942.1 | - | 3 - 4 | 667 | 57 | ||
| AK236014.1 | - | 3 - 4 | 462 | 57 | ||
| AK237883.1 | - | 3 - 4 | 568 | 57 | ||
| AK395073.1 | - | 3 - 4 | 488 | 57 | ||
| AK395180.1 | - | 4 - 5 | 636 | 57 | ||
| AK396654.1 | - | 3 - 4 | 488 | 57 | ||
| AK396827.1 | - | 4 - 5 | 564 | 57 | ||
| AK400586.1 | - | 3 - 4 | 655 | 57 | ||
| AK400598.1 | - | 3 - 4 | 545 | 57 | ||
More Information
Set Membership: |
|

TPM4
Genomic Map