Assay Details
| Gene Symbol: PTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214336.1 | NP_999501.1 | 2 - 3 | 359 | 70 | |
| XM_005673124.3 | XP_005673181.1 | 451 | 70 | |||
| XM_013985586.2 | XP_013841040.1 | 509 | 70 | |||
| GenBank mRNA | AK234367.1 | - | 2 - 3 | 368 | 70 | |
| AK347710.1 | - | 2 - 3 | 368 | 70 | ||
| AK400038.1 | - | 2 - 3 | 387 | 70 | ||
| AY609556.1 | - | 2 - 3 | 372 | 70 | ||
| D89546.1 | - | 2 - 3 | 359 | 70 | ||
More Information
Set Membership: |
|

PTN
Genomic Map