Assay Details
| Gene Symbol: PPP2CA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214366.1 | NP_999531.1 | 4 - 5 | 754 | 67 | |
| GenBank mRNA | AK235935.1 | - | 4 - 5 | 956 | 67 | |
| AY609733.1 | - | 4 - 5 | 804 | 67 | ||
| KU705627.1 | - | 4 - 5 | 575 | 67 | ||
| M20192.1 | - | 4 - 5 | 754 | 67 | ||
More Information
Set Membership: |
|

PPP2CA
Genomic Map