Assay Details
| Gene Symbol: LYZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214392.2 | NP_999557.2 | 3 - 4 | 439 | 87 | |
| GenBank mRNA | AK230563.1 | - | 3 - 4 | 87 | ||
| AK230612.1 | - | 3 - 4 | 87 | |||
| AK230919.1 | - | 3 - 4 | 87 | |||
| AK236844.1 | - | 3 - 4 | 87 | |||
| AK343904.1 | - | 3 - 4 | 87 | |||
| EW463673.2 | - | 3 - 4 | 401 | 87 | ||
| L14853.1 | - | 3 - 4 | 87 | |||
| U44435.1 | - | 3 - 4 | 87 | |||
More Information
Set Membership: |
|

LYZ
Genomic Map