Assay Details
| Gene Symbol: CYP2D25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214394.1 | NP_999559.1 | 1 - 2 | 251 | 101 | |
| GenBank mRNA | AK232448.1 | - | 1 - 2 | 267 | 101 | |
| AK232749.1 | - | 1 - 2 | 259 | 101 | ||
| AK346202.1 | - | 1 - 2 | 254 | 101 | ||
| AK398955.1 | - | 1 - 2 | 257 | 101 | ||
| AK400510.1 | - | 1 - 2 | 257 | 101 | ||
| KP687264.1 | - | 1 - 2 | 365 | 101 | ||
| Y16417.1 | - | 251 | 101 | |||
More Information
Set Membership: |
|

CYP2D25
Genomic Map