Assay Details
| Gene Symbol: CYP2D25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214394.1 | NP_999559.1 | 2 - 3 | 430 | 74 | |
| GenBank mRNA | AK232448.1 | - | 2 - 3 | 446 | 74 | |
| AK346280.1 | - | 2 - 3 | 434 | 74 | ||
| AK398955.1 | - | 2 - 3 | 436 | 74 | ||
| AK400510.1 | - | 2 - 3 | 436 | 74 | ||
| KP687264.1 | - | 2 - 3 | 544 | 74 | ||
| Y16417.1 | - | 430 | 74 | |||
More Information
Set Membership: |
|

CYP2D25
Genomic Map