Assay Details
| Gene Symbol: CYP2D25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214394.1 | NP_999559.1 | 3 - 4 | 584 | 80 | |
| GenBank mRNA | AK232448.1 | - | 3 - 4 | 600 | 80 | |
| AK232749.1 | - | 3 - 4 | 592 | 80 | ||
| AK346202.1 | - | 3 - 4 | 587 | 80 | ||
| AK346280.1 | - | 3 - 4 | 588 | 80 | ||
| AK394110.1 | - | 2 - 3 | 193 | 80 | ||
| AK398955.1 | - | 3 - 4 | 590 | 80 | ||
| AK400510.1 | - | 3 - 4 | 590 | 80 | ||
| D89502.1 | - | 3 - 4 | 549 | 80 | ||
| KP687264.1 | - | 3 - 4 | 698 | 80 | ||
| Y16417.1 | - | 584 | 80 | |||
More Information
Set Membership: |
|

CYP2D25
Genomic Map