Assay Details
| Gene Symbol: CYP2D25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214394.1 | NP_999559.1 | 4 - 5 | 738 | 64 | |
| GenBank mRNA | AK232448.1 | - | 4 - 5 | 754 | 64 | |
| AK400510.1 | - | 4 - 5 | 744 | 64 | ||
| D89502.1 | - | 4 - 5 | 703 | 64 | ||
| KP687264.1 | - | 4 - 5 | 852 | 64 | ||
| Y16417.1 | - | 738 | 64 | |||
More Information
Set Membership: |
|

CYP2D25
Genomic Map