Assay Details
| Gene Symbol: CYP2D25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214394.1 | NP_999559.1 | 5 - 6 | 916 | 72 | |
| GenBank mRNA | AK232448.1 | - | 5 - 6 | 932 | 72 | |
| AK232749.1 | - | 5 - 6 | 924 | 72 | ||
| AK346202.1 | - | 5 - 6 | 919 | 72 | ||
| AK346280.1 | - | 5 - 6 | 920 | 72 | ||
| AK394110.1 | - | 4 - 5 | 525 | 72 | ||
| AK398955.1 | - | 5 - 6 | 922 | 72 | ||
| AK400510.1 | - | 5 - 6 | 922 | 72 | ||
| D89502.1 | - | 5 - 6 | 881 | 72 | ||
| KP687264.1 | - | 5 - 6 | 1030 | 72 | ||
| Y16417.1 | - | 916 | 72 | |||
More Information
Set Membership: |
|

CYP2D25
Genomic Map