Assay Details
| Gene Symbol: CYP2D25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214394.1 | NP_999559.1 | 6 - 7 | 1057 | 67 | |
| GenBank mRNA | AK232448.1 | - | 6 - 7 | 1073 | 67 | |
| AK232749.1 | - | 6 - 7 | 1065 | 67 | ||
| AK346202.1 | - | 6 - 7 | 1060 | 67 | ||
| AK346280.1 | - | 6 - 7 | 1061 | 67 | ||
| AK394110.1 | - | 5 - 6 | 666 | 67 | ||
| AK398955.1 | - | 6 - 7 | 1063 | 67 | ||
| AK400510.1 | - | 6 - 7 | 1063 | 67 | ||
| D89502.1 | - | 6 - 7 | 1022 | 67 | ||
| KP687264.1 | - | 6 - 7 | 1171 | 67 | ||
| Y16417.1 | - | 1057 | 67 | |||
More Information
Set Membership: |
|

CYP2D25
Genomic Map