Assay Details
| Gene Symbol: SERPINA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001348942.1 | NP_001335871.1 | 4 - 5 | 1142 | 124 | |
| NM_214395.2 | NP_999560.2 | 4 - 5 | 1116 | 124 | ||
| GenBank mRNA | AK233443.1 | - | 4 - 5 | 1109 | 124 | |
| X88780.1 | - | 1079 | 124 | |||
More Information
Set Membership: |
|

SERPINA1
Genomic Map