Assay Details
| Gene Symbol: MMUT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214405.1 | NP_999570.1 | 12 - 13 | 2236 | 85 | |
| GenBank mRNA | AJ496801.1 | - | 12 - 13 | 2236 | 85 | |
| AK343989.1 | - | 12 - 13 | 2261 | 85 | ||
| AK389866.1 | - | 13 - 14 | 2234 | 85 | ||
| AK396479.1 | - | 12 - 13 | 2323 | 85 | ||
More Information
Set Membership: |
|

MMUT
Genomic Map