Assay Details
| Gene Symbol: MMUT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214405.1 | NP_999570.1 | 6 - 7 | 1440 | 75 | |
| GenBank mRNA | AJ496801.1 | - | 6 - 7 | 1440 | 75 | |
| AK343989.1 | - | 6 - 7 | 1465 | 75 | ||
| AK389866.1 | - | 7 - 8 | 1438 | 75 | ||
| AK396479.1 | - | 6 - 7 | 1527 | 75 | ||
More Information
Set Membership: |
|

MMUT
Genomic Map