Assay Details
| Gene Symbol: MMUT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214405.1 | NP_999570.1 | 7 - 8 | 1558 | 102 | |
| GenBank mRNA | AJ496801.1 | - | 7 - 8 | 1558 | 102 | |
| AK343989.1 | - | 7 - 8 | 1583 | 102 | ||
| AK389866.1 | - | 8 - 9 | 1556 | 102 | ||
| AK396479.1 | - | 7 - 8 | 1645 | 102 | ||
More Information
Set Membership: |
|

MMUT
Genomic Map