Assay Details
| Gene Symbol: MMUT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214405.1 | NP_999570.1 | 9 - 10 | 1792 | 81 | |
| GenBank mRNA | AJ496801.1 | - | 9 - 10 | 1792 | 81 | |
| AK343989.1 | - | 9 - 10 | 1817 | 81 | ||
| AK389866.1 | - | 10 - 11 | 1790 | 81 | ||
| AK396479.1 | - | 9 - 10 | 1879 | 81 | ||
More Information
Set Membership: |
|

MMUT
Genomic Map