Assay Details
Gene Symbol: CYP21A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214433.1 | NP_999598.1 | 1 - 2 | 206 | 73 | |
XM_005665806.3 | XP_005665863.1 | 360 | 73 | |||
XM_005665807.3 | XP_005665864.1 | 361 | 73 | |||
XM_021098321.1 | XP_020953980.1 | 366 | 73 | |||
XM_021098322.1 | XP_020953981.1 | 363 | 73 | |||
XM_021098323.1 | XP_020953982.1 | 364 | 73 | |||
XM_021098325.1 | XP_020953984.1 | 360 | 73 | |||
GenBank mRNA | AK343239.1 | - | 1 - 2 | 310 | 73 | |
AK343364.1 | - | 1 - 2 | 216 | 73 | ||
AK343420.1 | - | 1 - 2 | 252 | 73 | ||
AK343514.1 | - | 1 - 2 | 218 | 73 | ||
AK389399.1 | - | 1 - 2 | 218 | 73 | ||
AK389523.1 | - | 1 - 2 | 222 | 73 | ||
AK398798.1 | - | 1 - 2 | 216 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
