Assay Details
| Gene Symbol: SNAI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128439.2 | NP_001121911.2 | 2 | 536 | 117 | |
| GenBank mRNA | AB271932.1 | - | 2 | 405 | 117 | |
| DQ198158.1 | - | 1 | 151 | 117 | ||
| EU681192.1 | - | 2 | 471 | 117 | ||
More Information
Set Membership: |
|

SNAI2
Genomic Map