Assay Details
| Gene Symbol: TNFSF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025217.1 | NP_001020388.1 | 3 | 444 | 146 | |
| XM_021102256.1 | XP_020957915.1 | 1891 | 146 | |||
| GenBank mRNA | AY837783.1 | - | 3 | 444 | 146 | |
More Information
Set Membership: |
|

TNFSF4
Genomic Map