Assay Details
| Gene Symbol: SLA-DMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004039.1 | NP_001004039.1 | 1 - 2 | 141 | 93 | |
| NM_001113705.1 | NP_001107177.1 | 1 - 2 | 144 | 93 | ||
| GenBank mRNA | AB032169.1 | - | 1 - 2 | 141 | 93 | |
| AK350519.1 | - | 1 - 2 | 118 | 93 | ||
| AK352141.1 | - | 1 - 2 | 118 | 93 | ||
| AK398888.1 | - | 1 - 2 | 119 | 93 | ||
More Information
Set Membership: |
|

SLA-DMA
Genomic Map