Assay Details
| Gene Symbol: ADSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130733.1 | NP_001124205.1 | 5 - 6 | 674 | 63 | |
| GenBank mRNA | AK231893.1 | - | 5 - 6 | 704 | 63 | |
| AK231934.1 | - | 5 - 6 | 696 | 63 | ||
| EU779494.1 | - | 5 - 6 | 674 | 63 | ||
| GU249574.1 | - | 5 - 6 | 693 | 63 | ||
More Information
Set Membership: |
|

ADSL
Genomic Map