Assay Details
| Gene Symbol: ATIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130736.1 | NP_001124208.1 | 13 - 14 | 1371 | 63 | |
| GenBank mRNA | AK236628.1 | - | 13 - 14 | 1412 | 63 | |
| AK389977.1 | - | 13 - 14 | 1429 | 63 | ||
| EU779495.1 | - | 13 - 14 | 1371 | 63 | ||
More Information
Set Membership: |
|

ATIC
Genomic Map